Läs mer
2-(Dimethylamino)-2-phenylbutan-1-ol, TRC
Produktkod. 30373313
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30373313 10 mg
1 options
Produktkod. 30373313 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD50506510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Dimethylamino)-2-phenylbutan-1-ol
CAS 39068-94-5
Molekylformel C12H19NO
Kvantitet 10 mg
Synonym beta-(Dimethylamino)-beta-ethylbenzeneethanol, 2-(Dimethylamino)-2-phenylbutanol, 2-(N,N-Dimethylamino)-2-phenylbutanol
InChI formel InChI=1S/C12H19NO/c1-4-12(10-14,13(2)3)11-8-6-5-7-9-11/h5-9,14H,4,10H2,1-3H3
LEDER CCC(CO)(N(C)C)c1ccccc1
Molekylvikt (g/mol) 193.29