Läs mer
2-(Dimethylamino)ethyl Phenylacetate Hydrochloride, Mikromol™
Beskrivning
2-(Dimethylamino)ethyl Phenylacetate Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-(Dimethylamino)ethyl Phenylacetate Hydrochloride |
| CAS | 113079-81-5 |
| Analyt- eller komponentnamn | 2-(Dimethylamino)ethyl Phenylacetate Hydrochloride |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C12 H17 N O2 . Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Cyclopentolate Hydrochloride Imp. C (EP), Cyclopentolate Imp. C (EP), Benzeneacetic acid, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Benzeneacetic acid, 2-(dimethylamino)ethyl ester, hydrochloride (9CI), 2-(Dimethylamino)ethyl Phenylacetate Hydrochloride, Cyclopentolate Hydrochloride Imp. C (EP) as Hydrochloride, Cyclopentolate Hydrochloride Impurity C as Hydrochloride, Cyclopentolate Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C12H17NO2.ClH/c1-13(2)8-9-15-12(14)10-11-6-4-3-5-7-11;/h3-7H,8-10H2,1-2H3;1H |
| LEDER | Cl.CN(C)CCOC(=O)Cc1ccccc1 |
| IUPAC-namn | 2-(dimethylamino)ethyl 2-phenylacetate;hydrochloride |
| Visa mer |