Läs mer
2-(Dimethylamino)guanosine, TRC
Produktkod. 30012108
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012108 10 mg
30008224 5 mg
30009184 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012108 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD46085010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2140-67-2
Molekylformel C12 H17 N5 O5
Molekylvikt (g/mol) 311.29
Synonym N,N-Dimethylguanosine, 2-Dimethylamino-6-oxypurine Riboside;
IUPAC-namn 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-(dimethylamino)-1H-purin-6-one
LEDER CN(C)C1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1
Specifikationer
Kemiskt namn eller material 2-(Dimethylamino)guanosine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C12H17N5O5/c1-16(2)12-14-9-6(10(21)15-12)13-4-17(9)11-8(20)7(19)5(3-18)22-11/h4-5,7-8,11,18-20H,3H2,1-2H3,(H,14,15,21)/t5-,7-,8-,11-/m1/s1
Formel vikt 311.123
Procent renhet >95%