Läs mer
2-Dimethylaminoethanol Bitartrate, TRC
Produktkod. 30373213
Change view
Kvantitet:
250 g
Produktkod. Kvantitet
30373213 250 g
1 options
Produktkod. 30373213 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD471440250G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Dimethylaminoethanol Bitartrate
CAS 5988-51-2
Molekylformel C8H17NO7
Kvantitet 250 g
Synonym Deanol Bitartrate, Dimethylaminoethanol Bitartrate, Liparon, N,N-Dimethylethanolamine-tartaric Acid Salt, (2R,3R)-2,3-Dihydroxybutanedioate 2-(Dimethylamino)-ethanol, [R-(R*,R*)]-2,3-dihydroxybutanedioate 2-(Dimethylamino)-ethanol, 2-Dimethylaminoethanol Bitartrate, 2-Dimethylaminoethanol Tartrate
InChI formel InChI=1S/C4H11NO.C4H6O6/c1-5(2)3-4-6;5-1(3(7)8)2(6)4(9)10/h6H,3-4H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1
LEDER CN(C)CCO.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC-namn (2R,3R)-2,3-dihydroxybutanedioic acid;2-(dimethylamino)ethanol
Molekylvikt (g/mol) 239.22
Formel vikt 239.1005