Läs mer
2-Dimethylaminoethyl Benzilate, TRC
Produktkod. 30376700
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30376700 100 mg
1 options
Produktkod. 30376700 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD473730100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Dimethylaminoethyl Benzilate
CAS 968-46-7
Molekylformel C18H21NO3
Kvantitet 100 mg
Synonym 2-Dimethylaminoethyl Diphenylglycolate, Benzacine, CT 840, Hydroxydiphenine, Ifenti, Labotropin, Tropalpin, α-Hydroxy-α-phenylbenzeneacetic Acid 2-(Dimethylamino)ethyl Ester
InChI formel InChI=1S/C18H21NO3/c1-19(2)13-14-22-17(20)18(21,15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,21H,13-14H2,1-2H3
LEDER OC(C(OCCN(C)C)=O)(C1=CC=CC=C1)C2=CC=CC=C2
Molekylvikt (g/mol) 299.36
Formel vikt 299.1521