Läs mer
2-(Dimethylaminomethyl)-4-(2-aminoethylthiomethyl)thiazole, TRC
Produktkod. 17974775
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974775 500 mg
17901745 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17974775 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD461050500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78441-62-0
Molekylformel C9 H17 N3 S2
Molekylvikt (g/mol) 231.38
Synonym 2-Thiazolemethanamine, 4-[[(2-aminoethyl)thio]methyl]-N,N-dimethyl-, 4-[[(2-Aminoethyl)thio]methyl]-N,N-dimethyl-2-thiazolemethanamine, 2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethanamine, 2-(Dimethylaminomethyl)-4-(2-aminoethylthiomethyl)thiazole, Nizatidine Imp. D (EP), Nizatidine EP Impurity D
IUPAC-namn 2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethanamine
LEDER CN(C)Cc1nc(CSCCN)cs1
Specifikationer
Kemiskt namn eller material 2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethanamine
Kvantitet 500 mg
InChI formel InChI=1S/C9H17N3S2/c1-12(2)5-9-11-8(7-14-9)6-13-4-3-10/h7H,3-6,10H2,1-2H3
Formel vikt 231.0864