Läs mer
2-(Diphenylmethoxy)-N-methylethylamine, TRC
Produktkod. 30133059
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133059 1 g
30011188 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133059 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4916951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17471-10-2
Molekylformel C16 H19 N O
Molekylvikt (g/mol) 241.33
Synonym 2-(Diphenylmethoxy)-N-methylethanamine, Ethylamine, 2-(diphenylmethoxy)-N-methyl- (7CI,8CI), 2-(Diphenylmethoxy)-N-methylethylamine, FAR 8328V, N-Demethyldiphenhydramine, N-Desmethyldiphenhydramine, Nordiphenhydramine, Dimenhydrinate Imp. F (EP), Diphenhydramine Hydrochloride Imp. A (EP)
IUPAC-namn 2-benzhydryloxy-N-methylethanamine
LEDER CNCCOC(c1ccccc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Nordiphenhydramine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C16H19NO/c1-17-12-13-18-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15/h2-11,16-17H,12-13H2,1H3
Formel vikt 241.1467
Procent renhet >95%