missing translation for 'onlineSavingsMsg'
Learn More

2-Diphenylphosphino-1-naphthoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7026387
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
178176-80-2
C23H17O2P
356.36
MFCD02683090
WJCBDBBZIGUHOB-UHFFFAOYSA-N
2-diphenylphosphino-1-naphthoic acid, 2-diphenylphosphanyl naphthalene-1-carboxylic acid, acmc-20anx1, 2-diphenylphosphino-1-naphthalenecarboxylic acid, 2-diphenylphosphino-naphthalen-1-carboxylic acid, 1-naphthalenecarboxylicacid, 2-diphenylphosphino
10861198
2-diphenylphosphanylnaphthalene-1-carboxylic acid
OC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
178176-80-2
C23H17O2P
356.36
MFCD02683090
WJCBDBBZIGUHOB-UHFFFAOYSA-N
2-diphenylphosphino-1-naphthoic acid, 2-diphenylphosphanyl naphthalene-1-carboxylic acid, acmc-20anx1, 2-diphenylphosphino-1-naphthalenecarboxylic acid, 2-diphenylphosphino-naphthalen-1-carboxylic acid, 1-naphthalenecarboxylicacid, 2-diphenylphosphino
10861198
2-diphenylphosphanylnaphthalene-1-carboxylic acid
OC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190°C to 192°C
Quantity 1 g
Formula Weight 356.36
Percent Purity 97%
Chemical Name or Material 2-Diphenylphosphino-1-naphthoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'