Läs mer
2'-Epi-9,10-dihydroergotamine, TRC
Produktkod. 30007720
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30007720 25 mg -
30005562 2.5 mg 2.5mg
30003788 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007720 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58491025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5550-75-4
Molekylformel C33 H37 N5 O5
Molekylvikt (g/mol) 583.68
Synonym (2'β,5'α)-9,10-dihydro-12'-hydroxy-2'-methyl-5'-(phenylmethyl)ergotaman-3',6',18-trione, Aciergotamine, dihydro- (6CI,7CI), 2'-Epidihydroergotamine, Aci-Dihydroergotamine, (6aR,9R,10aR)-N-[(2S,5S,10aS,10bS)-5-benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10aoctahydroindolo[4,3-fg]quinoline-9-carboxamide, 2′-epi-9,10-dihydroergotamine, Dihydroergotamine Mesilate Imp. D (EP)
LEDER CN1C[C@@H](C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34)C(=O)N[C@@]5(C)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C5=O
Specifikationer
Kemiskt namn eller material 2'-Epidihydroergotamine
Kvantitet 25 mg
InChI formel InChI=1S/C33H37N5O5/c1-32(35-29(39)21-15-23-22-10-6-11-24-28(22)20(17-34-24)16-25(23)36(2)18-21)31(41)38-26(14-19-8-4-3-5-9-19)30(40)37-13-7-12-27(37)33(38,42)43-32/h3-6,8-11,17,21,23,25-27,34,42H,7,12-16,18H2,1-2H3,(H,35,39)/t21-,23-,25-,26+,27+,32+,33+/m1/s1
Formel vikt 583.2795