Läs mer
2-Ethenylbenzaldehyde, TRC
Produktkod. 30179999
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179999 100 mg
30180299 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179999 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE678170100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28272-96-0
Molekylformel C9H8O
Molekylvikt (g/mol) 132.16
Synonym 2-Formylstyrene, 2-Vinylbenzaldehyde, o-Formylstyrene, o-Vinylbenzaldehyde
LEDER C=Cc1ccccc1C=O
Specifikationer
Kemiskt namn eller material 2-Ethenylbenzaldehyde
Kvantitet 100 mg
InChI formel InChI=1S/C9H8O/c1-2-8-5-3-4-6-9(8)7-10/h2-7H,1H2
Formel vikt 132.0575