Läs mer
2'-Ethoxyacetophenone, 97+%, Thermo Scientific Chemicals
Produktkod. 11440631
Change view
Kvantitet:
10 g
50 g
Förpackningsstorlek:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11440631 10 g 10g
11450631 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11440631 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B20113.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2142-67-8
Molekylformel C10H12O2
Molekylvikt (g/mol) 164.204
MDL-nummer MFCD00017262
InChI-nyckel TVGMOUGXQYQZOL-UHFFFAOYSA-N
Synonym 2'-ethoxyacetophenone, 1-2-ethoxyphenyl ethanone, ethanone, 1-2-ethoxyphenyl, 2-acetylphenetole, 1-2-ethoxyphenyl ethan-1-one, 1-acetyl-2-ethoxybenzene, o-ethoxyacetophenone, pubchem13191, acmc-1bt18, ethanone,2-ethoxy-1-phenyl
PubChem CID 578482
IUPAC-namn 1-(2-ethoxyphenyl)ethanone
LEDER CCOC1=CC=CC=C1C(=O)C
Specifikationer
Kemiskt namn eller material 2'-Ethoxyacetophenone
Smältpunkt 38°C to 42°C
Densitet 1
Kokpunkt 244°C
Flampunkt >110°C (230°F)
Kvantitet 10 g
Formel vikt 164.2
Procent renhet 98%