missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl acetate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7021345
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy
111-15-9
C6H12O3
132.16
MFCD00009251
SVONRAPFKPVNKG-UHFFFAOYSA-N
ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
8095
2-ethoxyethyl acetate
CCOCCOC(C)=O
This item is not returnable. View return policy
111-15-9
C6H12O3
132.16
MFCD00009251
SVONRAPFKPVNKG-UHFFFAOYSA-N
ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
8095
2-ethoxyethyl acetate
CCOCCOC(C)=O

2-Ethoxyethyl acetate is used as a solvent dissolve resin, leather, ink, for the formulation of paints, lacquers and varnishes for industrial use. It is also used to study the field evaluation of a passive badge for measuring 2-ethoxyethyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxyethyl acetate is used as a solvent dissolve resin, leather, ink, for the formulation of paints, lacquers and varnishes for industrial use. It is also used to study the field evaluation of a passive badge for measuring 2-ethoxyethyl acetate.

Solubility
Soluble in water(230g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point -62°C
Density 0.975
Boiling Point 155°C to 157°C
Flash Point 52°C (126°F)
Odor Fruit-like
Linear Formula CH3CO2CH2CH2OCH2CH3
Refractive Index 1.407
Quantity 500 mL
UN Number UN1172
Beilstein 1748677
Merck Index 14,3751
Solubility Information Soluble in water(230g/L).
Formula Weight 132.16
Percent Purity ≥98%
Chemical Name or Material 2-Ethoxyethyl acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'