Läs mer
2-Ethoxyhexa-3,5-diene, TRC
Produktkod. 30387518
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30387518 250 mg
30385568 100 mg
30381879 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30387518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE567520250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56752-55-7
Molekylformel C8H14O
Molekylvikt (g/mol) 126.2
Synonym 5-Ethoxy-1,3-hexadiene
IUPAC-namn (3E)-5-ethoxyhexa-1,3-diene
LEDER CCOC(C)\C=C\C=C
Specifikationer
Kemiskt namn eller material 2-Ethoxyhexa-3,5-diene
Kvantitet 250 mg
InChI formel InChI=1S/C8H14O/c1-4-6-7-8(3)9-5-2/h4,6-8H,1,5H2,2-3H3/b7-6+
Formel vikt 126.1045