missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7077475
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
848243-23-2
C13H20BNO3
249.12
MFCD07366721
GRXNEZGBXVIORK-UHFFFAOYSA-N
2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-ethoxypyridine-3-boronic acid pinacol ester, 2-ethoxypyridin-3-ylboronic acid pinacol ester, 2-ethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem16583, 2-ethoxypyridin-3-yl boronic acid pinacol ester
17750191
2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CCOC1=C(C=CC=N1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
848243-23-2
C13H20BNO3
249.12
MFCD07366721
GRXNEZGBXVIORK-UHFFFAOYSA-N
2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-ethoxypyridine-3-boronic acid pinacol ester, 2-ethoxypyridin-3-ylboronic acid pinacol ester, 2-ethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem16583, 2-ethoxypyridin-3-yl boronic acid pinacol ester
17750191
2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CCOC1=C(C=CC=N1)B1OC(C)(C)C(C)(C)O1
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 249.12
Percent Purity 95%
Chemical Name or Material 2-Ethoxypyridine-3-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Ethoxypyridine-3-boronic acid pinacol ester, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'