Learn More
2-ethyl-2-methyloctanoic acid, TRC
Product Code. 30346963
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30346963 50 mg 50mg
1 options
Product Code. 30346963 Supplier Toronto Research Chemicals Supplier No. TRCB44118050MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-ethyl-2-methyloctanoic acid
CAS 31199-56-1
Molecular Formula C11H22O2
Quantity 50 mg
InChI Formula InChI=1S/C11H22O2/c1-4-6-7-8-9-11(3,5-2)10(12)13/h4-9H2,1-3H3,(H,12,13)
SMILES CCCCCCC(C)(CC)C(=O)O
IUPAC Name 2-ethyl-2-methyloctanoic acid
Molecular Weight (g/mol) 186.29
Formula Weight 186.162