Läs mer
2-Ethyl-2-methylpentanoic Acid, TRC
Produktkod. 30136048
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136048 5 mg
30012104 50 mg
17919554 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136048 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9242905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5343-52-2
Molekylformel C8 H16 O2
Molekylvikt (g/mol) 144.21
Synonym 2-Ethyl-2-methylpentanoic acid, Valeric acid, 2-ethyl-2-methyl- (6CI,8CI), 2-Ethyl-2-methylpentanoic acid, 2-Methyl-2-ethylpentanoic acid, NSC 466, α-Ethyl-α-methylvaleric acid, α-Methyl-α-ethylvaleric acid, (2RS)-2-Ethyl-2-methylpentanoic Acid
IUPAC-namn 2-ethyl-2-methylpentanoic acid
LEDER CCCC(C)(CC)C(=O)O
Specifikationer
Kemiskt namn eller material 2-Ethyl-2-methylpentanoic Acid
Kvantitet 5 mg
InChI formel InChI=1S/C8H16O2/c1-4-6-8(3,5-2)7(9)10/h4-6H2,1-3H3,(H,9,10)
Formel vikt 144.115