Läs mer
2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate), Mikromol™
Beskrivning
2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate)
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate) |
| CAS | 80866-90-6 |
| Analyt- eller komponentnamn | 2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate) |
| Frakt skick | Room Temperature |
| Molekylformel | C11 H14 N2 O2 . H2 O |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Primidone Imp. A (EP), Primidone USP Related Compound A, Primidone USP RC A, 2-Ethyl-2-phenylpropane-diamide Hydrate, Primidone Imp. A (EP) as Hydrate, Ethylphenylmalonamide Hydrate, Primidone Related Compound A as Monohydrate, Primidone Impurity A as Hydrate |
| InChI formel | InChI=1S/C11H14N2O2.H2O/c1-2-11(9(12)14,10(13)15)8-6-4-3-5-7-8;/h3-7H,2H2,1H3,(H2,12,14)(H2,13,15);1H2 |
| LEDER | O.CCC(C(=O)N)(C(=O)N)c1ccccc1 |
| IUPAC-namn | 2-ethyl-2-phenylpropanediamide;hydrate |
| Visa mer |