Läs mer
2-Ethyl-6-methylpyrazine, TRC
Produktkod. 30133841
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133841 50 mg
30132913 100 mg
17913375 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133841 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB43820850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13925-03-6
Molekylformel C7H10N2
Molekylvikt (g/mol) 122.17
Synonym Pyrazine, 2-ethyl-6-methyl-, 2-Ethyl-6-methylpyrazine, 2-Methyl-6-ethylpyrazine, 6-Methyl-2-ethylpyrazine
IUPAC-namn 2-ethyl-6-methylpyrazine
LEDER CCC1=NC(=CN=C1)C
Specifikationer
Kemiskt namn eller material 2-Ethyl-6-methylpyrazine
Kvantitet 50 mg
InChI formel InChI=1S/C7H10N2/c1-3-7-5-8-4-6(2)9-7/h4-5H,3H2,1-2H3
Formel vikt 122.0844