Learn More
2-Ethyl-N,N-dimethylhexan-1-amine, 98%, BLD Pharm™
Product Code. 30948552
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948552 250 mg 250mg
30936027 100 mg 100mg
30946048 5 g 5g
30947222 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30948552 Supplier BLD Pharm Supplier No. BD142161250mg
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 28056-87-3
Molecular Formula C10H23N
MDL Number MFCD00126935
Quantity 250 mg
SMILES CCCCC(CC)CN(C)C
Molecular Weight (g/mol) 157.30
Percent Purity 98%
Chemical Name or Material 2-Ethyl-N,N-dimethylhexan-1-amine