Learn More
2-(Ethylamino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Etilefrone Hydrochloride), Mikromol™
Product Code. 30049741
Change view
Click to view available options
Kvantitet:
25 mg
Product Code. Kvantitet
30049741 25 mg
1 options
Product Code. 30049741 Supplier Mikromol Supplier No. MM0035.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(Ethylamino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Etilefrone Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Etilefrone Hydrochloride
CAS 22510-04-9
Analyt- eller komponentnamn Etilefrone Hydrochloride
Frakt skick Room Temperature
Molekylformel C10 H13 N O2 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Ethanone, 2-(ethylamino)-1-(3-hydroxyphenyl)-, hydrochloride (1:1), Acetophenone, 2-(ethylamino)-3'-hydroxy-, hydrochloride (8CI), Ethanone, 2-(ethylamino)-1-(3-hydroxyphenyl)-, hydrochloride (9CI), 2-(Ethylamino)-1-(3-hydroxyphenyl)ethanone hydrochloride, Etilefrone hydrochloride
InChI formel InChI=1S/C10H13NO2.ClH/c1-2-11-7-10(13)8-4-3-5-9(12)6-8;/h3-6,11-12H,2,7H2,1H3;1H
LEDER Cl.CCNCC(=O)c1cccc(O)c1
IUPAC-namn 2-(ethylamino)-1-(3-hydroxyphenyl)ethanone;hydrochloride
Molekylvikt (g/mol) 215.6767
Kvantitet 25 mg
Formel vikt 215.0713 g/mol