missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7077845
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
22927-13-5
C9H10O
134.18
MFCD02261766
NTWBHJYRDKBGBR-UHFFFAOYSA-N
benzaldehyde, 2-ethyl, o-ethylbenzaldehyde, 2-ethyl benzaldehyde, 2-ethyl-benzaldehyde, ethylbenzaldehyde, 2-ethyl-benzaldehyd, 6-ethyl benzaldehyde, pubchem3074, acmc-20a49p
123406
2-ethylbenzaldehyde
CCC1=CC=CC=C1C=O
This item is not returnable. View return policy
22927-13-5
C9H10O
134.18
MFCD02261766
NTWBHJYRDKBGBR-UHFFFAOYSA-N
benzaldehyde, 2-ethyl, o-ethylbenzaldehyde, 2-ethyl benzaldehyde, 2-ethyl-benzaldehyde, ethylbenzaldehyde, 2-ethyl-benzaldehyd, 6-ethyl benzaldehyde, pubchem3074, acmc-20a49p
123406
2-ethylbenzaldehyde
CCC1=CC=CC=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0200g/mL
Boiling Point 209.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Refractive Index 1.538
Quantity 5 g
Specific Gravity 1.02
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 2-Ethylbenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'