Läs mer
(2-Ethylbenzofuran-3-yl)(4-hydroxyphenyl)methanone (Benzarone), Mikromol™
Produktkod. 30051615
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051615 25 mg
30050541 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051615 Leverantör Mikromol Leverantörsnummer MM0456.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2-Ethylbenzofuran-3-yl)(4-hydroxyphenyl)methanone (Benzarone)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzarone
CAS 1477-19-6
Analyt- eller komponentnamn Benzarone
Frakt skick Room Temperature
Molekylformel C17 H14 O3
Certifikat och efterlevnad ISO 17025
Synonym Benzbromarone Imp. C (EP), (2-Ethyl-3-benzofuranyl)(4-hydroxyphenyl)methanone, (2-Ethylbenzofuran-3-yl)(4-hydroxyphenyl)methanone, 2-Ethyl-3-(4-hydroxybenzoyl)benzofuran, 2-Ethyl-3-(p-hydroxybenzoyl)benzofuran, 2-Ethyl-3-benzofuranyl p-hydroxyphenyl ketone, 2-Ethyl-4'-hydroxy-3-benzoylbenzofuran, Benzaron, Benzarone, Fagivil, Fragivil, Fragivix, L 2179-Labaz, L 2197, NSC 82134, Benzbromarone Impurity C
InChI formel InChI=1S/C17H14O3/c1-2-14-16(13-5-3-4-6-15(13)20-14)17(19)11-7-9-12(18)10-8-11/h3-10,18H,2H2,1H3
LEDER CCc1oc2ccccc2c1C(=O)c3ccc(O)cc3
IUPAC-namn (2-ethyl-1-benzofuran-3-yl)-(4-hydroxyphenyl)methanone
Molekylvikt (g/mol) 266.29
Kvantitet 25 mg
Formel vikt 266.0943 g/mol