Läs mer
2-Ethylhexyl 4-Nitrobenzoate, TRC
Produktkod. 30373939
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373939 250 mg
30375964 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30373939 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE918880250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16397-70-9
Molekylformel C15H21NO4
Molekylvikt (g/mol) 279.33
Synonym 2-Ethylhexyl p-Nitrobenzoate, 4-Nitro-benzoic Acid 2-Ethylhexyl Ester
IUPAC-namn 2-ethylhexyl 4-nitrobenzoate
LEDER O=C(OCC(CC)CCCC)C1=CC=C(C=C1)N(=O)=O
Specifikationer
Kemiskt namn eller material 2-Ethylhexyl 4-Nitrobenzoate
Kvantitet 250 mg
InChI formel InChI=1S/C15H21NO4/c1-3-5-6-12(4-2)11-20-15(17)13-7-9-14(10-8-13)16(18)19/h7-10,12H,3-6,11H2,1-2H3
Formel vikt 279.15