Läs mer
2-Ethylhexyl Diphenyl Phosphate (90%), TRC
Produktkod. 17918184
Change view
Kvantitet:
250 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17918184 250 mg
17950085 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17918184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE917980250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1241-94-7
Molekylformel C20 H27 O4 P
Molekylvikt (g/mol) 362.4
Synonym Phosphoric acid, 2-ethylhexyl diphenyl ester (6CI, 7CI, 8CI, 9CI, ACI), 2-Ethylhexyl diphenyl phosphate, Diphenyl 2-ethylhexyl phosphate, Disflamoll DPO, EHDPP, Octicizer, Phosflex 342, Phosflex 362, Santicizer 141, 2-Ethylhexyldiphenyl Phosphate
IUPAC-namn 2-ethylhexyl diphenyl phosphate
LEDER CCCCC(CC)COP(=O)(Oc1ccccc1)Oc2ccccc2
Specifikationer
Kemiskt namn eller material 2-Ethylhexyl diphenyl phosphate
Kvantitet 250 mg
InChI formel InChI=1S/C20H27O4P/c1-3-5-12-18(4-2)17-22-25(21,23-19-13-8-6-9-14-19)24-20-15-10-7-11-16-20/h6-11,13-16,18H,3-5,12,17H2,1-2H3
Formel vikt 362.1647