Läs mer
2-Ethylpentanoic Acid, TRC
Produktkod. 30008048
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008048 1 g
30004153 250 mg
18020510 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008048 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8180001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20225-24-5
Molekylformel C7 H14 O2
Molekylvikt (g/mol) 130.18
Synonym (2RS)-2-Ethylpentanoic Acid, Valproic Acid Imp. B (EP)
IUPAC-namn 2-ethylpentanoic acid
LEDER CCCC(CC)C(=O)O
Specifikationer
Kemiskt namn eller material (2RS)-2-Ethylpentanoic Acid
Kvantitet 1 g
InChI formel InChI=1S/C7H14O2/c1-3-5-6(4-2)7(8)9/h6H,3-5H2,1-2H3,(H,8,9)
Formel vikt 130.0994