Läs mer
2-(Ethylthio)-1H-benzimidazole, TRC
Produktkod. 30135658
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135658 1 g
17904185 500 mg
17962245 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135658 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9268851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14610-11-8
Molekylformel C9H10N2S
Molekylvikt (g/mol) 178.25
Synonym 2-(Ethylsulfanyl)benzimidazole, 2-(Ethylthio)benzimidazole, 2-Mercaptoethylbenzimidazole, Bemithyl
IUPAC-namn 2-ethylsulfanyl-1H-benzimidazole
LEDER CCSc1nc2ccccc2[nH]1
Specifikationer
Kemiskt namn eller material 2-(Ethylthio)-1H-benzimidazole
Kvantitet 1 g
InChI formel InChI=1S/C9H10N2S/c1-2-12-9-10-7-5-3-4-6-8(7)11-9/h3-6H,2H2,1H3,(H,10,11)
Formel vikt 178.0565