Learn More
2-Fluoro-3-formylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Catalog Number p-7054279
Click to view available options
Quantity:
250 mg
1 g
1112209-40-1
C13H16BFO3
250.076
MFCD17015836
IFIDGFJKIAHLAQ-UHFFFAOYSA-N
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-fluoro-3-formylphenylboronic acid pinacol ester, 2-fluoro-3-formylphenylboronic acid, pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde
53217345
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C=O)F
1112209-40-1
C13H16BFO3
250.076
MFCD17015836
IFIDGFJKIAHLAQ-UHFFFAOYSA-N
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-fluoro-3-formylphenylboronic acid pinacol ester, 2-fluoro-3-formylphenylboronic acid, pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde
53217345
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C=O)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Fluoro-3-formylbenzeneboronic acid pinacol ester is used as a intermediate in pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 250.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3-formylbenzeneboronic acid pinacol ester