missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitrotoluene, 98%, Thermo Scientific Chemicals

Catalog Number p-7054769
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
437-86-5
C7H6FNO2
155.128
MFCD03412242
NBCNUIXYBLFJMI-UHFFFAOYSA-N
2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
13470957
2-fluoro-1-methyl-3-nitrobenzene
CC1=C(C(=CC=C1)[N+](=O)[O-])F
This item is not returnable. View return policy
437-86-5
C7H6FNO2
155.128
MFCD03412242
NBCNUIXYBLFJMI-UHFFFAOYSA-N
2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
13470957
2-fluoro-1-methyl-3-nitrobenzene
CC1=C(C(=CC=C1)[N+](=O)[O-])F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 19°C (decomposition)
Color Colorless to Yellow
Density 1.276 g/mL
Boiling Point 111°C (12 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.529
Quantity 25 g
UN Number UN2810
Beilstein 2442082
Formula Weight 155.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-nitrotoluene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'