missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog Number p-3759054
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
115029-22-6
C8H3F4O2
207.10
MFCD00040980
XVEAMDNSCPPPCP-UHFFFAOYSA-M
2-fluoro-3-trifluoromethyl benzoic acid, 2-fluoro-3-trifluoromethyl benzoicacid, alpha,alpha,alpha,2-tetrafluoro-m-toluic acid, 2-fluoro-3-trifluoromethyl-benzoic acid, benzoic acid, 2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethylbenzioc acid, pubchem2741, acmc-1boz3, ksc489q1l, rarechem al bo 0625
518002
2-fluoro-3-(trifluoromethyl)benzoic acid
[O-]C(=O)C1=CC=CC(=C1F)C(F)(F)F
This item is not returnable. View return policy
115029-22-6
C8H3F4O2
207.10
MFCD00040980
XVEAMDNSCPPPCP-UHFFFAOYSA-M
2-fluoro-3-trifluoromethyl benzoic acid, 2-fluoro-3-trifluoromethyl benzoicacid, alpha,alpha,alpha,2-tetrafluoro-m-toluic acid, 2-fluoro-3-trifluoromethyl-benzoic acid, benzoic acid, 2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethylbenzioc acid, pubchem2741, acmc-1boz3, ksc489q1l, rarechem al bo 0625
518002
2-fluoro-3-(trifluoromethyl)benzoic acid
[O-]C(=O)C1=CC=CC(=C1F)C(F)(F)F
CAS Min % 97.5
CAS Max % 100.0
Melting Point 126.0°C to 128.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H3(CF3)CO2H
Quantity 1 g
Formula Weight 208.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-3-(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'