missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-4'-hydroxybenzophenone, 98%

255.75 SEK - 732.22 SEK

Chemical Identifiers

CAS 101969-75-9
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.211
MDL Number MFCD01075694
InChI Key FHFDJJWFCDGBEM-UHFFFAOYSA-N
Synonym 2-fluoro-4'-hydroxybenzophenone, 2-fluorophenyl 4-hydroxyphenyl methanone, 4-2-fluorobenzoyl phenol, 2-fluorophenyl-4-hydroxyphenyl methanone, 2-fluorophenyl 4-hydroxyphenyl ketone, acmc-20anv8, 2-fluoro-4/'-hydroxybenzophenone, 2-fluorophenyl 4-hydroxy-phenyl methanone, methanone, 2-fluorophenyl 4-hydroxyphenyl
PubChem CID 2783235
IUPAC Name (2-fluorophenyl)-(4-hydroxyphenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)O)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358276 SDS View Documents Alfa Aesar™
L15494.03
1g 255.75 SEK
1g View alternative products
Save Up to 
»
EU Stock 2
Add to basket
 
11368276 SDS View Documents Alfa Aesar™
L15494.06
5g 732.22 SEK
5g View alternative products
Save Up to 
»
EU Stock 3
Add to basket
 
Specifications

Chemical Identifiers

101969-75-9
216.211
FHFDJJWFCDGBEM-UHFFFAOYSA-N
2783235
C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)O)F
C13H9FO2
MFCD01075694
2-fluoro-4'-hydroxybenzophenone, 2-fluorophenyl 4-hydroxyphenyl methanone, 4-2-fluorobenzoyl phenol, 2-fluorophenyl-4-hydroxyphenyl methanone, 2-fluorophenyl 4-hydroxyphenyl ketone, acmc-20anv8, 2-fluoro-4/'-hydroxybenzophenone, 2-fluorophenyl 4-hydroxy-phenyl methanone, methanone, 2-fluorophenyl 4-hydroxyphenyl
(2-fluorophenyl)-(4-hydroxyphenyl)methanone

Specifications

1g
115°C to 118°C
C13H9FO2
2-fluoro-4'-hydroxybenzophenone, 2-fluorophenyl 4-hydroxyphenyl methanone, 4-2-fluorobenzoyl phenol, 2-fluorophenyl-4-hydroxyphenyl methanone, 2-fluorophenyl 4-hydroxyphenyl ketone, acmc-20anv8, 2-fluoro-4/'-hydroxybenzophenone, 2-fluorophenyl 4-hydroxy-phenyl methanone, methanone, 2-fluorophenyl 4-hydroxyphenyl
C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)O)F
216.211
216.21
2-Fluoro-4'-hydroxybenzophenone
101969-75-9
MFCD01075694
FHFDJJWFCDGBEM-UHFFFAOYSA-N
(2-fluorophenyl)-(4-hydroxyphenyl)methanone
2783235
98%