missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-4-iodo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65877.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449966

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-4-iodo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
395.22
MFCD28098833

Specifications

2-Fluoro-4-iodo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
1g
95%
MFCD28098833
Light Sensitive
Solid
395.22
Videos
SDS