missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodoaniline, 98%, Thermo Scientific™

Catalog Number p-3211688
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
29632-74-4
C6H5FIN
237.01
MFCD00011738
CUMTUBVTKOYYOU-UHFFFAOYSA-N
2-fluoro-4-iodo-phenylamine, benzenamine, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenamine, 2-fluoro-4-iodo aniline, 2-fluoro-4-iodo-aniline, 2-fluoro-4-iodophenylamine, zlchem 543, pubchem1532, 2-fluoro4-iodoaniline, 2-fluoro-4-iodaniline
185694
2-fluoro-4-iodoaniline
C1=CC(=C(C=C1I)F)N
This item is not returnable. View return policy
29632-74-4
C6H5FIN
237.01
MFCD00011738
CUMTUBVTKOYYOU-UHFFFAOYSA-N
2-fluoro-4-iodo-phenylamine, benzenamine, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenamine, 2-fluoro-4-iodo aniline, 2-fluoro-4-iodo-aniline, 2-fluoro-4-iodophenylamine, zlchem 543, pubchem1532, 2-fluoro4-iodoaniline, 2-fluoro-4-iodaniline
185694
2-fluoro-4-iodoaniline
C1=CC(=C(C=C1I)F)N
CAS Min % 97.5
CAS Max % 100.0
Melting Point 51°C to 57°C
Color White to Brown
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H3(F)NH2
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 237.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-4-iodoaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'