missing translation for 'onlineSavingsMsg'
Läs mer

2'-Fluoro-4'-methylacetophenone, 97%, Thermo Scientific™

Produktkod. 11322639
Click to view available options
Kvantitet:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11322639

Brand: Thermo Scientific Alfa Aesar H26695.06

Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Detta Thermo Scientific varumärkesprodukt var ursprungligen en del av Alfa Aesar produktportfölj. Viss dokumentation och etikettinformation kan referera till det äldre varumärket. Originalet Alfa Aesar produkt-/artikelkod eller SKU-referens har inte ändrats som en del av varumärkesövergången till Thermo Scientific .

Applications
2′-Fluoro-4′-methylacetophenone is used as an intermediate in pharmaceuticals.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29427-48-3
Molekylformel C9H9FO
Molekylvikt (g/mol) 152.17
MDL-nummer MFCD03094331
InChI-nyckel IBABHBXTLZNGAV-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-methylphenyl ethanone, 2'-fluoro-4'-methylacetophenone, 1-2-fluoro-4-methylphenyl ethan-1-one, ethanone, 1-2-fluoro-4-methylphenyl, acmc-1cjnp, 4-acetyl-3-fluorotoluene, 2-fluoro-4-methylacetophenone, 1-acetyl-2-fluoro-4-methylbenzene, ethanone,1-2-fluoro-4-methylphenyl
PubChem CID 2778484
IUPAC-namn 1-(2-fluoro-4-methylphenyl)ethanone
LEDER CC(=O)C1=C(F)C=C(C)C=C1

Specifications

Kemiskt namn eller material 2'-Fluoro-4'-methylacetophenone
Kvantitet 5 g
Formel vikt 152.17
Procent renhet 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.