Läs mer
2-Fluoro-4-methylbenzeneboronic acid, 97%, Thermo Scientific™
Produktkod. 15480018
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15480018 5 g 5g
15470018 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15480018 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H53265.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 170981-26-7
Molekylformel C7H8BFO2
Molekylvikt (g/mol) 153.947
MDL-nummer MFCD05664239
InChI-nyckel LIXXGOMAGHXIMP-UHFFFAOYSA-N
Synonym 2-fluoro-4-methylphenyl boronic acid, 2-fluoro-4-methylbenzeneboronic acid, 2-fluoro-4-methyl-phenyl boronic acid, boronic acid, 2-fluoro-4-methylphenyl, 2-fluoro-p-tolylboronic acid, 4-methyl-2-fluorophenylboronic acid, boronic acid, b-2-fluoro-4-methylphenyl, pubchem23775, ksc489k8t, 2-fluoro-4methylphenylboronic acid
PubChem CID 2783132
IUPAC-namn (2-fluoro-4-methylphenyl)boronic acid
LEDER B(C1=C(C=C(C=C1)C)F)(O)O
Specifikationer
Kemiskt namn eller material 2-Fluoro-4-methylbenzeneboronic acid
Smältpunkt 228°C to 233°C
Kvantitet 5 g
Formel vikt 153.95
Procent renhet 97%