missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitroaniline, 98%, ACROS Organics™

Brand:  Acros Organics 457600250

Additional Details : CAS Number : 369-35-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15575256

  • 840.00 SEK / 25g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

369-35-7
156.11
2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
2-fluoro-4-nitroaniline
C6H5FN2O2
LETNCFZQCNCACQ-UHFFFAOYSA-N
101254
C1=CC(=C(C=C1[N+](=O)[O-])F)N

Specifications

2-Fluoro-4-nitroaniline
122.0°C to 130.0°C
369-35-7
C6H5FN2O2
2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
C1=CC(=C(C=C1[N+](=O)[O-])F)N
156.11
156.11
Authentic
25g
97.5% min. (GC)
FC6H3(NO2)NH2
LETNCFZQCNCACQ-UHFFFAOYSA-N
2-fluoro-4-nitroaniline
101254
98%
Videos
SDS
Documents