missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitroaniline, 98%, Thermo Scientific™

Catalog Number p-7077687
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
369-35-7
C6H5FN2O2
156.11
LETNCFZQCNCACQ-UHFFFAOYSA-N
2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
101254
2-fluoro-4-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])F)N
This item is not returnable. View return policy
369-35-7
C6H5FN2O2
156.11
LETNCFZQCNCACQ-UHFFFAOYSA-N
2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
101254
2-fluoro-4-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])F)N
Melting Point 122.0°C to 130.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H3(NO2)NH2
Quantity 5 g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'