missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-3759052
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
126917-10-0
C8H3ClF4O
226.56
MFCD00061155
OOAHPLWBUUTFMV-UHFFFAOYSA-N
2-fluoro-4-trifluoromethyl benzoyl chloride, 2-fluoro-4-trifluoromethyl benzoylchloride, benzoyl chloride, 2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-benzoyl chloride, benzoyl chloride,2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-1-benzenecarbonyl chloride, alpha,alpha,alpha,2-tetrafluoro-p-toluoyl chloride, acmc-1c5cr, 2-fluoro-4-trifluoromethyl-benzoylchloride, 4-trifluoromethyl-2-fluorobenzoyl chloride
145580
2-fluoro-4-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C=C1C(F)(F)F)F)C(=O)Cl
This item is not returnable. View return policy
126917-10-0
C8H3ClF4O
226.56
MFCD00061155
OOAHPLWBUUTFMV-UHFFFAOYSA-N
2-fluoro-4-trifluoromethyl benzoyl chloride, 2-fluoro-4-trifluoromethyl benzoylchloride, benzoyl chloride, 2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-benzoyl chloride, benzoyl chloride,2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-1-benzenecarbonyl chloride, alpha,alpha,alpha,2-tetrafluoro-p-toluoyl chloride, acmc-1c5cr, 2-fluoro-4-trifluoromethyl-benzoylchloride, 4-trifluoromethyl-2-fluorobenzoyl chloride
145580
2-fluoro-4-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C=C1C(F)(F)F)F)C(=O)Cl
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.5000g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CF3)COCl
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.5
Formula Weight 226.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'