Learn More
2-Fluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.
Solubility
Miscible with water, alcohol, acetonitrile and N,N-dimethylformamide
Notes
Incompatible with oxidizing agents and bases.
2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.
Solubility
Miscible with water, alcohol, acetonitrile and N,N-dimethylformamide
Notes
Incompatible with oxidizing agents and bases.
Specifications
Boiling Point | 113°C |
Quantity | 1 g |
Solubility Information | Miscible with water,alcohol,acetonitrile and N,N-dimethylformamide |
Formula Weight | 165.09 |
Percent Purity | 97% |
Chemical Name or Material | 2-Fluoro-4-(trifluoromethyl)pyridine |