missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methyl-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog Number p-7052422
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1373920-66-1
C8H7F4N
193.15
MFCD22201049
SCCCZPNCTRGYDF-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl aniline
86277650
2-fluoro-5-methyl-4-(trifluoromethyl)aniline
CC1=CC(N)=C(F)C=C1C(F)(F)F
This item is not returnable. View return policy
1373920-66-1
C8H7F4N
193.15
MFCD22201049
SCCCZPNCTRGYDF-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl aniline
86277650
2-fluoro-5-methyl-4-(trifluoromethyl)aniline
CC1=CC(N)=C(F)C=C1C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2810
Formula Weight 193.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-methyl-4-(trifluoromethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'