missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™

Catalog Number 15448955
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-16-0
C9H8F4O
208.156
MFCD19687197
YSXVMRQKBSXFCF-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl benzyl alcohol, 2-fluoro-5-methyl-4-trifluoromethyl phenyl methanol
66523486
[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]methanol
CC1=CC(=C(C=C1C(F)(F)F)F)CO
This item is not returnable. View return policy
1323966-16-0
C9H8F4O
208.156
MFCD19687197
YSXVMRQKBSXFCF-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl benzyl alcohol, 2-fluoro-5-methyl-4-trifluoromethyl phenyl methanol
66523486
[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]methanol
CC1=CC(=C(C=C1C(F)(F)F)F)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 208.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'