missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Fluoro-5-(trifluoromethoxy)benzyl bromide is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-5-(trifluoromethoxy)benzyl bromide is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Fluoro-5-(trifluoromethoxy)benzyl bromide is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Refractive Index | 1.466 |
Quantity | 1 g |
UN Number | UN3265 |
Solubility Information | Difficult to mix. |
Formula Weight | 273.02 |
Percent Purity | 97% |
Chemical Name or Material | 2-Fluoro-5-(trifluoromethoxy)benzyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'