missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride is used to describe a straight or branched chain alkyl.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride is used to describe a straight or branched chain alkyl.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Refractive Index | 1.478 |
Quantity | 250 mg |
UN Number | UN3265 |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 262.61 |
Percent Purity | 97% |
Chemical Name or Material | 2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'