missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number p-7054528
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1599432-41-3
C13H18BFO3
252.092
CSXSULPDIWBLHT-UHFFFAOYSA-N
2-2-fluoro-6-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-6-methoxybenzeneboronic acid pinacol ester, 2-fluoro-6-methoxyphenylboronic acid pinacol ester, 2-fluoro-6-methoxybenzeneboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronic acid 2,3-dimethyl-2,3-butanediyl
72221147
2-(2-fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)OC
This item is not returnable. View return policy
1599432-41-3
C13H18BFO3
252.092
CSXSULPDIWBLHT-UHFFFAOYSA-N
2-2-fluoro-6-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-6-methoxybenzeneboronic acid pinacol ester, 2-fluoro-6-methoxyphenylboronic acid pinacol ester, 2-fluoro-6-methoxybenzeneboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronic acid 2,3-dimethyl-2,3-butanediyl
72221147
2-(2-fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 252.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-6-methoxybenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'