missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-phenoxybenzaldehyde, 96%, Thermo Scientific™

Catalog Number 11434870
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
902836-68-4
C13H9FO2
216.211
MFCD03428585
AUPPVEMCDSSXCW-UHFFFAOYSA-N
2-fluoro-6-phenoxy-benzaldehyde
24721181
2-fluoro-6-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C=O
This item is not returnable. View return policy
902836-68-4
C13H9FO2
216.211
MFCD03428585
AUPPVEMCDSSXCW-UHFFFAOYSA-N
2-fluoro-6-phenoxy-benzaldehyde
24721181
2-fluoro-6-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 36°C to 39°C
Quantity 250 mg
Formula Weight 216.21
Percent Purity 96%
Chemical Name or Material 2-Fluoro-6-phenoxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'