missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-6-(trifluoromethyl)benzeneboronic acid, 98%

Brand:  Alfa Aesar™ H64146.03

Additional Details : CAS Number : 313545-34-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498726

  • 683.27 SEK / 1g


Estimated Shipment date
from Supplier 12-12-2019
Add to basket

Specifications

Chemical Identifiers

313545-34-5
207.919
TXBRJTKFAYXKMV-UHFFFAOYSA-N
16217168
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O
C7H5BF4O2
MFCD07781239
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenylboron&, 2-fluoro-6-trifluoromethylphenyl boronic acid
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid

Specifications

115°C to 119°C
2-Fluoro-6-(trifluoromethyl)benzeneboronic acid
C7H5BF4O2
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenylboron&, 2-fluoro-6-trifluoromethylphenyl boronic acid
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O
207.919
207.92
98%
1g
313545-34-5
MFCD07781239
TXBRJTKFAYXKMV-UHFFFAOYSA-N
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid
16217168
Solid
Videos
SDS Documents