missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-fluoro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline-11-carboxylic acid, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Quantity | 250 mg |
Formula Weight | 259.28 |
Percent Purity | 97% |
Chemical Name or Material | 2-fluoro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline-11-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'