Läs mer
2-Fluoro Atorvastatin Sodium Salt, TRC
Produktkod. 30404308
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30404308 100 mg
1 options
Produktkod. 30404308 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF587900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Fluoro Atorvastatin Sodium Salt
CAS 1346598-86-4
Molekylformel C33H34FN2NaO5
Kvantitet 100 mg
Synonym (βR,ΔR)-2-(2-Fluorophenyl)-β,Δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic Acid Sodium Salt
InChI formel InChI=1S/C33H35FN2O5.Na/c1-21(2)31-30(33(41)35-23-13-7-4-8-14-23)29(22-11-5-3-6-12-22)32(26-15-9-10-16-27(26)34)36(31)18-17-24(37)19-25(38)20-28(39)40;/h3-16,21,24-25,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);/q;+1/p-1/t24-,25-;/m1./s1
LEDER O=C(NC1=CC=CC=C1)C2=C(C(C)C)N(CC[C@@H](O)C[C@@H](O)CC(O[Na])=O)C(C3=C(F)C=CC=C3)=C2C4=CC=CC=C4
Molekylvikt (g/mol) 580.62