Learn More
2-Fluorophenacyl bromide, 98%, ACROS Organics™
Brand:  Acros Organics 446670050
Additional Details : CAS Number : 655-15-2 Weight : 0.05500kg Transport : UN number : 3265 Chem class : 8 Pack group : III
Quantity 5g
 View more versions of this product
Product Code. 12065400
  • 883.00 SEK / 5g
Estimated Shipment: 16-08-2021
Specifications
Chemical Identifiers
655-15-2
217.04
2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone
2-bromo-1-(2-fluorophenyl)ethanone
C8H6BrFO
QDNWNJSLWKHNTM-UHFFFAOYSA-N
2737449
C1=CC=C(C(=C1)C(=O)CBr)F
Specifications
2-Fluorophenacyl bromide
5g
97.5% min. (GC)
2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone
C1=CC=C(C(=C1)C(=O)CBr)F
217.04
217.04
61.0°C to 63.0°C (0.1 Torr)
655-15-2
C8H6BrFO
QDNWNJSLWKHNTM-UHFFFAOYSA-N
2-bromo-1-(2-fluorophenyl)ethanone
2737449
98%
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 2-Fluorophenacyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.