Läs mer
2-Formamidoacetamide, TRC
Produktkod. 30405828
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30405828 500 mg
1 options
Produktkod. 30405828 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF735565500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Formamidoacetamide
CAS 4238-57-7
Molekylformel C3 H6 N2 O2
Kvantitet 500 mg
Synonym 2-(Formylamino)acetamide, Formylglycinamide, N-Formylglycinamide, N-Formylglycine amide, NSC 29551, Nα-Formylglycinamide
InChI formel InChI=1S/C3H6N2O2/c4-3(7)1-5-2-6/h2H,1H2,(H2,4,7)(H,5,6)
LEDER NC(=O)CNC=O
IUPAC-namn 2-formamidoacetamide
Molekylvikt (g/mol) 102.092
Formel vikt 102.043