Läs mer
2-Formyl-3-oxopropanoic Acid Ethyl Ester, TRC
Produktkod. 30407946
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30407946 1 g
1 options
Produktkod. 30407946 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF7008951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Formyl-3-oxopropanoic Acid Ethyl Ester
CAS 80370-42-9
Molekylformel C6H8O4
Kvantitet 1 g
Synonym (Ethoxycarbonyl)malonaldehyde, 2-Formyl-3-oxo-propionic Acid Ethyl Ester, Ethyl 2,2-Diformylacetate, Ethyl 2-Formyl-3-oxopropanoate
InChI formel InChI=1S/C6H8O4/c1-2-10-6(9)5(3-7)4-8/h3-5H,2H2,1H3
LEDER CCOC(=O)C(C=O)C=O
IUPAC-namn ethyl 2-formyl-3-oxopropanoate
Molekylvikt (g/mol) 144.13
Formel vikt 144.0423